missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-GPR109B/HM74, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human GPR109B was used as the immunogen.
Antigen GPR109B/HM74
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224488 Novus Biologicals Canada
NBP224488
0.1mg Each for $255.80
NBP224488SS Novus Biologicals Canada
NBP224488SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
GPR109B/HM74
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 86% in rhesus monkey and 73% homologous in bovine and human REG3G.
100μg in 200μL PBS containing 0.2% gelatin
A portion of amino acids 1-50 of human GPR109B was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
8843
0.5mg/mL