Please login to your online account to display your discounted pricing

anti-GPIHBP1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11041537SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11041537T

  • / Each
 

Description & Specifications

Specifications

Antigen GPIHBP1
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.95mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation Tris-glycine, 150mM NaCl
Host Species Rabbit
Immunogen A synthetic peptide corresponding to an internal region [within residues 100-170] of mouse GPIHBP1. [Swiss-Prot No. Q9D1N2]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 338328
Test Specificity Mouse. Rat and Bovine reactivity reported in scientific literature (PMID: 24735886).
Target Species Bovine
Target Species Murine
Target Species Rat