Please login to your online account to display your discounted pricing

anti-GPIHBP1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to an internal region [within residues 100-170] of mouse GPIHBP1. [Swiss-Prot No. Q9D1N2]
Antigen GPIHBP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11041537

 
novus biologicals canada
NB11041537
Unlabeled 0.1mL Each for $329.00

NB11041537T

 
novus biologicals canada
NB11041537SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to an internal region [within residues 100-170] of mouse GPIHBP1. [Swiss-Prot No. Q9D1N2]
Antigen GPIHBP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Mouse. Rat and Bovine reactivity reported in scientific literature (PMID: 24735886).
Gene ID (Entrez) 338328
Formulation Tris-glycine, 150mM NaCl
Concentration 0.95mg/mL