missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-GOLM1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen This antibody was raised against GOLPH 2
Antigen GOLM1
Monoclonal or Polyclonal Polyclonal
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227212 Novus Biologicals Canada
NBP227212
0.1mg Each for $255.80
NBP227212SS Novus Biologicals Canada
NBP227212SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
This antibody was raised against GOLPH 2
Polyclonal
Primary
ELISA,Immunohistochemistry (Paraffin)
100μg antibody in 200μL PBS containing 0.05% BSA.
IgG
GOLM1
Purified
RUO
51280
0.5mg/mL