Please login to your online account to display your discounted pricing

anti-GOLM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227212SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227212SS

  • / Each
 

Description & Specifications

Specifications

Antigen GOLM1
Applications ELISA
Applications Immunohistochemistry (Paraffin)
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Host Species Rabbit
Immunogen This antibody was raised against GOLPH 2
Isotype IgG
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51280
Target Species Human