Please login to your online account to display your discounted pricing

anti-GOLM1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227212

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227212

  • / Each
 

Description & Specifications

Specifications

Isotype IgG
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 51280
Conjugate Unlabeled
Primary or Secondary Primary
Cross Reactivity Human
Concentration 0.5mg/mL
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Host Species Rabbit
Immunogen This antibody was raised against GOLPH 2
Regulatory Status RUO
Antigen GOLM1
Applications ELISA
Applications Immunohistochemistry (Paraffin)
Format Purified
Quantity 0.1mg