Please login to your online account to display your discounted pricing

anti-Podoplanin, Clone: 8.1.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001015

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001015

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Primary or Secondary Primary
Test Specificity Mouse. Does not cross-react with human.
Formulation Tris-glycine, 150mM NaCl
Gene ID (Entrez) 10630
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Applications Electron Microscopy
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Murine thymic stromal cell lines
Antigen Podoplanin
Clone 8.1.1
Format Purified
Quantity 0.1mL
Cross Reactivity Mouse
Host Species Golden Syrian Hamster
Isotype IgG
Regulatory Status RUO