Please login to your online account to display your discounted pricing

anti-GNPDA1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224557

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224557

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P46926
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene ID (Entrez) 10007
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, horse, dog, pig, cow, opossum, chicken, chimpanzee, rat, mouse and rhesus monkey, 93% homologous in zebrafish, platypus and salmon, 87% homologous in pufferfish and 75% homologous in Xenopus.
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Formulation 100μg in 200μL PBS containing 0.05% BSA
Format Affinity Purified
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Primate
Immunogen A portion of amino acids 250-289 of human GNPDA1 was used as the immunogen.
Antigen GNPDA1
Regulatory Status RUO
Primary or Secondary Primary
Host Species Rabbit
Quantity 0.1mg