missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-GNPDA1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 250-289 of human GNPDA1 was used as the immunogen.
Antigen GNPDA1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224557 Novus Biologicals Canada
NBP224557
0.1mg Each for $257.41
NBP224557SS Novus Biologicals Canada
NBP224557SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
GNPDA1
Affinity Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, horse, dog, pig, cow, opossum, chicken, chimpanzee, rat, mouse and rhesus monkey, 93% homologous in zebrafish, platypus and salmon, 87% homologous in pufferfish and 75% homologous in Xenopus.
10007
0.5mg/mL
A portion of amino acids 250-289 of human GNPDA1 was used as the immunogen.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
P46926
100μg in 200μL PBS containing 0.05% BSA