Please login to your online account to display your discounted pricing

anti-GM-CSF, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-75 of human CSF2 was used as the immunogen. 2138
Antigen GM-CSF
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227161

 
novus biologicals canada
NBP227161
Unlabeled 0.1mg Each for $255.80

NBP227161SS

 
novus biologicals canada
NBP227161SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-75 of human CSF2 was used as the immunogen. 2138
Antigen GM-CSF
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rat and chimpanzee, 94% homologous in monkey.
Gene Accession No. P04141
Gene ID (Entrez) 1437
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL