Please login to your online account to display your discounted pricing

anti-Glutathione Reductase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224940SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224940SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Format Purified
Test Specificity This antibody is reactive to Human, S. cerevisiae.
Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.05% BSA
Applications Western Blotting
Quantity 0.025mg
Gene ID (Entrez) 2936
Cross Reactivity Human
Cross Reactivity Yeast
Host Species Rabbit
Regulatory Status RUO
Immunogen Glutathione Reductase from bakers yeast (S. cerevisiae)
Monoclonal or Polyclonal Polyclonal
Antigen Glutathione Reductase