Please login to your online account to display your discounted pricing

anti-Glutathione Reductase, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224940

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224940

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Purified
Monoclonal or Polyclonal Polyclonal
Test Specificity This antibody is reactive to Human, S. cerevisiae.
Formulation 100μg in 200μL PBS containing 0.05% BSA
Applications Western Blotting
Quantity 0.1mg
Gene ID (Entrez) 2936
Immunogen Glutathione Reductase from bakers yeast (S. cerevisiae)
Cross Reactivity Human
Cross Reactivity Yeast
Antigen Glutathione Reductase
Regulatory Status RUO
Host Species Rabbit