Please login to your online account to display your discounted pricing

anti-Glutathione Reductase, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen Glutathione Reductase from bakers yeast (S. cerevisiae)
Antigen Glutathione Reductase
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224940

 
novus biologicals canada
NBP224940
Unlabeled 0.1mg Each for $257.41

NBP224940SS

 
novus biologicals canada
NBP224940SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Glutathione Reductase from bakers yeast (S. cerevisiae)
Antigen Glutathione Reductase
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity This antibody is reactive to Human, S. cerevisiae.
Gene ID (Entrez) 2936
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL