Please login to your online account to display your discounted pricing

anti-Glutaredoxin 1/GLRX1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081753SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081753T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P35754
Primary or Secondary Primary
Gene ID (Entrez) 2745
Conjugate Unlabeled
Formulation Tris-citrate/phosphate, pH 7-8
Test Specificity Human. Predicted to react with mouse, rat, rabbit, porcine, bovine based on 100% sequence homology. Immunogen sequence has 91% homology to Zebrafish.
Quantity 0.025mL
Concentration 1mg/mL
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Regulatory Status RUO
Immunogen Synthetic peptide made to an internal portion of human Glutaredoxin 1 (within residues 40-100). [Swiss-Prot No. P35754]
Host Species Rabbit
Cross Reactivity Human
Applications Western Blotting
Antigen Glutaredoxin 1/GLRX1