Please login to your online account to display your discounted pricing

anti-Glutaredoxin 1/GLRX1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081753

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081753

  • / Each
 

Description & Specifications

Specifications

Antigen Glutaredoxin 1/GLRX1
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. P35754
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human Glutaredoxin 1 (within residues 40-100). [Swiss-Prot No. P35754]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 2745
Test Specificity Human. Predicted to react with mouse, rat, rabbit, porcine, bovine based on 100% sequence homology. Immunogen sequence has 91% homology to Zebrafish.
Target Species Human