Please login to your online account to display your discounted pricing

anti-GLUT9, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP105054

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP105054

  • / Each
 

Description & Specifications

Specifications

Antigen GLUT9
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. Q9NRM0
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human GLUT9 (within residues 500-550). [Swiss-Prot No. Q9NRM0]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 56606
Target Species Human
Target Species Murine