missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Glut4, Polyclonal, Novus Biologicals™

$144.63 - $384.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human Glucose Transporter GLUT4 protein (between residues 480-509) [UniProt P14672]
Antigen Glut4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP149533 Novus Biologicals Canada
NBP149533
0.1mL Each for $384.00
NBP149533SS Novus Biologicals Canada
NBP149533SS
0.025mL Each for $144.63
Specifications Description & Specifications

Specifications

Rabbit
Glut4
Affinity Purified
RUO
Human and mouse. Predicted to react with rat based on 100% sequence homology.
6517
1.14mg/mL
A synthetic peptide made to a C-terminal portion of the human Glucose Transporter GLUT4 protein (between residues 480-509) [UniProt P14672]
Polyclonal
Primary
Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
P14672
PBS, 30% glycerol