Please login to your online account to display your discounted pricing

anti-Glut2, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human GLUT2 protein (between residues 50-150) [UniProt P11168]
Antigen Glut2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222218SS

 
novus biologicals canada
NBP222218SS
Unlabeled 0.025mL Each for $139.00

NBP222218

 
novus biologicals canada
NBP222218
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human GLUT2 protein (between residues 50-150) [UniProt P11168]
Antigen Glut2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human and mouse. Immunogen sequence has 84% homology to bovine and 78% homology to rat.
Gene Accession No. P11168
Gene ID (Entrez) 6514
Formulation PBS
Concentration 1.0mg/mL