Please login to your online account to display your discounted pricing

anti-Glut1, Polyclonal, Novus Biologicals™

$164.00 - $164.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal region of the human GLUT1 protein (between residues 1-100). [Swiss-Prot No. P11166]
Antigen Glut1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11039113T

 
novus biologicals canada
NB11039113SS
Unlabeled 0.025mL Each for $164.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal region of the human GLUT1 protein (between residues 1-100). [Swiss-Prot No. P11166]
Antigen Glut1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Test Specificity Reacts with human and mouse. 100% sequence identity with primate protein, 93% sequence identity with rat, cow, and rabbit proteins.
Gene Accession No. P11166
Gene ID (Entrez) 6513
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL