Please login to your online account to display your discounted pricing

GLUT-1 Polyclonal Antibody, Invitrogen™


GLUT-1 Antibody, PA5-16793, from Invitrogen™. Species Reactivity: Human, Rat; Applications: Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot.

Manufacturer: pierce biotechnology inc  PA516793

Catalog No. PIPA516793

  • / Each
 

Description & Specifications

Specifications

Formulation 500μL purified antibody (0.2mg/mL) in PBS with 0.2% BSA and 15mM sodium azide; pH 7.6
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Symbols SLC2A1
Gene ID (Entrez) 24778, 6513
Gene Accession No. P11166, P11167
Gene Alias GLUT-1, glucose transporter type 1, erythrocyte/brain, hepG2 glucose transporter, solute carrier family 2, facilitated glucose transporter member 1
Species Reactivity Human, Rat
Storage Requirements Store at 4° C
Immunogen A synthetic peptide derived from C-terminal of human GLUT-1
Quantity 500μL
Format Purified
Antigen GLUT-1
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Rat
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Isotype IgG
Regulatory Status RUO

GLUT-1 Antibody, PA5-16793, from Invitrogen. Species Reactivity: Human, Rat; Applications: Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot.

Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus, and breast. GLUT-1 immunoreactivity in some cancers, including trans carcinoma of the urinary bladder, has been associated with aggressive behavior.
Host Species: Rabbit
Cross Reactivity: Human, Rat
Isotype: IgG
Immunogen: A synthetic peptide derived from C-terminal of human GLUT-1
Formulation: 500μL purified antibody (0.2mg/mL) in PBS with 0.2% BSA and 15mM sodium azide; pH 7.6
Synonyms: GLUT-1, glucose transporter type 1, erythrocyte/brain, hepG2 glucose transporter, solute carrier family 2, facilitated glucose transporter member 1
Uniprot Accession: P11166, P11167
Entrez GeneID: 24778, 6513
Gene Symbol: SLC2A1
PA5-16793 targets GLUT-1 in IHC (P), WB, and IF applications and shows reactivity with Human and Rat samples. The PA5-16793 immunogen is a synthetic peptide derived from C-terminal of human GLUT-1.
References:
Test: ICC, Species: Hu, Reference: Human monocyte-derived hemangioma-like endothelial cells: evidence from an in vitro study. (2008 Jul-Aug) Cardiovasc Pathol, Vol: 17, 4, 212-8, Jia J, Huang X, Zhang WF, Zhao YF; Pubmed ID: 18402811