GluR2 Rabbit anti-Mouse, Rat, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma AB1768I
Catalog No. AB1768IMI
Description
Description
This GluR2 antibody is validated for use in western blotting and immunohistochemistry for the detection of the GluR2 protein.
Specifications
Specifications
GluR-2
Polyclonal
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Rabbit
Affinity Purified
Primary
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Gria2; Glur2
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of rat GluR2.
25 μg
NP_058957
2-8°C for 12 months