Glucose Transporter GLUT-3 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier:  MilliporeSigma AB1344
Catalog No. AB1344MI
Description
Description
Anti-Glucose Transporter GLUT-3 Antibody, C-terminus is an antibody against Glucose Transporter GLUT-3 for use in ELISA and western blotting.
Specifications
Specifications
Glucose Transporter GLUT-3
Polyclonal
Liquid rabbit antiserum containing 0.05% sodium azide.
Synthetic peptide corresponding to the C-terminus of mouse Glut-3 coupled to KLH. Product sold sold as AG617
Primary
Mouse, Rat
Serum
ELISA, Western Blot
Unconjugated
Rabbit
50 μL
NP_035531.3
-20°C to -80°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles