Please login to your online account to display your discounted pricing

anti-Glucagon Oligoclonal, Thermo Scientific™

Glucagon Oligoclonal antibody 710320 from Thermo Scientific™ specifically detects Glucagon in Human and Mouse samples, and it is validated for Western Blot

$424.20 - $424.20

Products
Catalog Number Mfr. No. Price Quantity    

PI710320

 
pierce biotechnology inc
710320
Each for $424.20
Description & Specifications

Glucagon Oligoclonal antibody 710320 from Thermo Scientific™ specifically detects Glucagon in Human and Mouse samples, and it is validated for Western Blot

Although pathogenic bacteria constitute only a small portion of bacterial species, infectious diseases caused by pathogenic bacteria affect millions of people worldwide. Diarrheal diseases, many of which are bacterial, are the second leading cause of death worldwide, claiming an estimated 5 million people annually. Furthermore, an estimated 3 million people die of tuberculosis annually, 500,000 people die of pertussis, and 25,000 die of typhoid. The ability of bacteria to utilize various food sources is a major factor behind their pervasiveness. Their vast numbers lead to rapid evolutionary changes resulting in emerging diseases as well as antibiotic-resistant strains of well known diseases. Examples of these are multi-drug resistant tuberculosis and methicillin-resistant Staphylococcus aureus.
Host Species: Rabbit
Cross Reactivity: Human, Mouse
Isotype: IgG
Clone: 24HCLC
Immunogen: Peptide corresponding to amino acids 98–118 of human GLP-1
Formulation: 100μg purified Antibody (0.5mg/mL) in PBS, with 0.09% sodium azide;
Synonyms: glicentin-related polypeptide, glucagon-like peptide 1, glucagon-like peptide 2, GCG, GLP1, GLP2, GRPP
Uniprot Accession: P01275, P55095
Gene Symbol: GCG
Entrez GeneID: 14526, 2641
MA5-18186 detects Chlamydia trachomatis LPS in Bacteria samples. MA5-18186 has been successfully used in ELISA procedures. MA5-18186 The immunogen is Chlamydia sp. native protein.