Please login to your online account to display your discounted pricing

anti-GLP-1R, Polyclonal, Novus Biologicals™

$192.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal extracellular portion of the human GLP1R protein (between residues 100-200) [UniProt P43220]
Antigen GLP-1R
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NLS1205001M

 
novus biologicals canada
NLS1205001ML
Unlabeled 0.025mL Each for $192.00

NLS1205

 
novus biologicals canada
NLS1205
Unlabeled 0.05mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal extracellular portion of the human GLP1R protein (between residues 100-200) [UniProt P43220]
Antigen GLP-1R
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Test Specificity Human and mouse. The immunogen used for this antibody has 100/88% homology/identity with the rat protein.
Gene Accession No. P43220
Gene ID (Entrez) 2740
Formulation PBS
Concentration 1.0mg/mL