Please login to your online account to display your discounted pricing

anti-GLP-1R, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NLS1205

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NLS1205

  • / Each
 

Description & Specifications

Specifications

Test Specificity Human and mouse. The immunogen used for this antibody has 100/88% homology/identity with the rat protein.
Formulation PBS
Gene ID (Entrez) 2740
Concentration 1.0mg/mL
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Primary or Secondary Primary
Gene Accession No. P43220
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal extracellular portion of the human GLP1R protein (between residues 100-200) [UniProt P43220]
Antigen GLP-1R
Quantity 0.05mL
Format Affinity Purified
Regulatory Status RUO