Please login to your online account to display your discounted pricing

anti-GLP-1R, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NLS1205001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NLS1205001M

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P43220
Concentration 1.0mg/mL
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Conjugate Unlabeled
Test Specificity Human and mouse. The immunogen used for this antibody has 100/88% homology/identity with the rat protein.
Gene ID (Entrez) 2740
Formulation PBS
Regulatory Status RUO
Quantity 0.025mL
Host Species Rabbit
Format Affinity Purified
Monoclonal or Polyclonal Polyclonal
Immunogen A synthetic peptide made to an N-terminal extracellular portion of the human GLP1R protein (between residues 100-200) [UniProt P43220]
Antigen GLP-1R