Please login to your online account to display your discounted pricing

anti-GLI-3, Polyclonal, Novus Biologicals™

$109.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human Gli3 protein (between residues 50-120) [UniProt P10071]
Antigen GLI-3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP229627

 
novus biologicals canada
NBP229627
Unlabeled 0.1mg Each for $259.02

NBP229627SS

 
novus biologicals canada
NBP229627SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of human Gli3 protein (between residues 50-120) [UniProt P10071]
Antigen GLI-3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human and mouse. Immunogen has 84% homology to Xenopus and 85% homology to chicken.
Gene Accession No. P10071
Gene ID (Entrez) 2737
Formulation PBS
Concentration 1.0mg/mL