Please login to your online account to display your discounted pricing

anti-GLI-2, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human Gli2 protein (between residues 300-400) [UniProt P10070]
Antigen GLI-2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223602SS

 
novus biologicals canada
NBP223602SS
Unlabeled 0.025mL Each for $139.00

NBP223602

 
novus biologicals canada
NBP223602
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human Gli2 protein (between residues 300-400) [UniProt P10070]
Antigen GLI-2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Test Specificity Human and mouse. Predicted to react with rat and hamster based on sequence homology.
Gene Accession No. P10070
Gene ID (Entrez) 2736
Formulation PBS
Concentration 1.0mg/mL