missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-GLI-1, Polyclonal, Novus Biologicals™

$164.00 - $384.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made toward the C-terminus portion of the human Gli1 protein (between residues 800-850) [UniProt P08151]
Antigen GLI-1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB600600 Novus Biologicals Canada
NB600600
0.1mL Each for $384.00
NB600600SS Novus Biologicals Canada
NB600600SS
0.025mL Each for $164.00
Specifications Description & Specifications

Specifications

Rabbit
GLI-1
Affinity Purified
RUO
This is directed against Gli1 from mouse brain. No reaction occurs with human or mouse Gli2 or Gli3.
2735
1.05mg/mL
A synthetic peptide made toward the C-terminus portion of the human Gli1 protein (between residues 800-850) [UniProt P08151]
Polyclonal
Primary
Chromatin Immunoprecipitation,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Western Blot
P08151
PBS, 30% glycerol