Please login to your online account to display your discounted pricing

anti-GLI-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600600

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600600

  • / Each
 

Description & Specifications

Specifications

Formulation PBS, 30% glycerol
Applications Chromatin Immunoprecipitation
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A synthetic peptide made toward the C-terminus portion of the human Gli1 protein (between residues 800-850) [UniProt P08151]
Antigen GLI-1
Gene ID (Entrez) 2735
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Gene Accession No. P08151
Primary or Secondary Primary
Test Specificity This is directed against Gli1 from mouse brain. No reaction occurs with human or mouse Gli2 or Gli3.
Concentration 1.05mg/mL
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mL
Regulatory Status RUO
Format Affinity Purified