Please login to your online account to display your discounted pricing

anti-GFP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600310SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600310SS

  • / Each
 

Description & Specifications

Specifications

Formulation Tris-citrate/phosphate, pH 7-8
Concentration 0.232mg/mL
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Quantity 0.025mL
Cross Reactivity All
Primary or Secondary Primary
Regulatory Status RUO
Test Specificity Reacts against all variants of recombinant Aequorea GFP. However, there is little to no reactivity with Renilla GFP.
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Conjugate Unlabeled
Immunogen Highly purified native GFP Aequorea victoria.
Host Species Rabbit
Antigen GFP