Please login to your online account to display your discounted pricing

anti-GFP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600310

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600310

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Primary or Secondary Primary
Test Specificity Reacts against all variants of recombinant Aequorea GFP. However, there is little to no reactivity with Renilla GFP.
Formulation Tris-citrate/phosphate, pH 7-8
Antigen GFP
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Concentration 0.232mg/mL
Cross Reactivity All
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Quantity 0.05mL
Host Species Rabbit
Format Affinity Purified
Immunogen Highly purified native GFP Aequorea victoria.