Please login to your online account to display your discounted pricing

anti-GFP, Polyclonal, Novus Biologicals™

$144.63 - $329.00

Specifications

Host Species Rabbit
Immunogen Highly purified native GFP Aequorea victoria.
Antigen GFP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600310

 
novus biologicals canada
NB600310
Unlabeled 0.05mL Each for $329.00

NB600310SS

 
novus biologicals canada
NB600310SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Highly purified native GFP Aequorea victoria.
Antigen GFP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Test Specificity Reacts against all variants of recombinant Aequorea GFP. However, there is little to no reactivity with Renilla GFP.
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 0.232mg/mL