Please login to your online account to display your discounted pricing

anti-GDNF Polyclonal, Thermo Scientific™

GDNF Polyclonal antibody PA1-18359 from Thermo Scientific™ specifically detects GDNF in Human, Mouse and Rat samples, and it is validated for Western Blot

$543.75 - $543.75

Specifications

Antigen GDNF
Cross Reactivity Human
Format Serum
Host Species Sheep
Immunogen Recombinant human GDNF.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-611

 
pierce biotechnology inc
PA118359
Unlabeled 100μL Each for $543.75

01-670-619

 
pierce biotechnology inc
PA118367
Unlabeled 100μL Each for $543.75
Description & Specifications

Specifications

Antigen GDNF
Cross Reactivity Human
Format Serum
Host Species Sheep
Immunogen Recombinant human GDNF.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

GDNF Polyclonal antibody PA1-18359 from Thermo Scientific™ specifically detects GDNF in Human, Mouse and Rat samples, and it is validated for Western Blot

GDNF is a glycosylated, disulfide-bonded homodimer molecule. It was first discovered as a potent survival factor for midbrain dopaminergic neurons and was then shown to rescue these neurons in animal models of Parkinson's disease. GDNF is about 100 times more efficient survival factor for spinal motor neurons than the neurotrophins. GDNF is a neurotrophic factor that enhances survival and morphological differentiation of dopaminergic neurons and increases their high-affinity dopamine uptake.
Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: Recombinant human GDNF.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Glial cell line-derived neurotrophic factor, Astrocyte-derived trophic factor 1, ATF1, ATF2, HFB1-GDNF
Uniprot Accession: P39905, P48540, Q07731
Gene Symbol: GDNF
Entrez GeneID: 14573, 25453, 2668
PA1-18359 detects GDNF in Human, Mouse and Rat samples. PA1-18359 has been successfully used in Western Blot procedures. PA1-18359 The immunogen is Recombinant human GDNF..