Please login to your online account to display your discounted pricing

anti-GBP1, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 500-550 of human GBP1 was used as the immunogen for this antibody.
Antigen GBP1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP203972

 
novus biologicals canada
NBP203972
Unlabeled 0.1mg Each for $307.00

NBP203972SS

 
novus biologicals canada
NBP203972SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 500-550 of human GBP1 was used as the immunogen for this antibody.
Antigen GBP1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Reacts with Human and Mouse. The amino acid sequence used as immunogen has 100% identity in human, 95% in rat and 79% in mouse.
Gene Accession No. NP_002044.2
Gene ID (Entrez) 2633
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA