Please login to your online account to display your discounted pricing

anti-GBL, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224705

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224705

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 64223
Concentration 0.5mg/mL
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Conjugate Unlabeled
Immunogen A portion of amino acids 150-200 of human LST8 was used as the immunogen.
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, opossum and platypus, 92% homologous in chicken and Xenopus, 88% homologous in zebrafish and 78% homologous in cow, horse, dog and rhesus monkey.
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Antigen GBL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q9BVC4
Quantity 0.1mg
Regulatory Status RUO
Applications Western Blotting
Isotype IgG
Format Purified
Host Species Rabbit