missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-GBL, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 150-200 of human LST8 was used as the immunogen.
Antigen GBL
Monoclonal or Polyclonal Polyclonal
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224705 Novus Biologicals Canada
NBP224705
0.1mg Each for $255.80
NBP224705SS Novus Biologicals Canada
NBP224705SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
A portion of amino acids 150-200 of human LST8 was used as the immunogen.
Polyclonal
Primary
Western Blot
Q9BVC4
100μg in 200μL PBS containing 0.2% gelatin
IgG
GBL
Purified
RUO
The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, opossum and platypus, 92% homologous in chicken and Xenopus, 88% homologous in zebrafish and 78% homologous in cow, horse, dog and rhesus monkey.
64223
0.5mg/mL