Please login to your online account to display your discounted pricing

anti-GBL, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224705SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224705SS

  • / Each
 

Description & Specifications

Specifications

Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, opossum and platypus, 92% homologous in chicken and Xenopus, 88% homologous in zebrafish and 78% homologous in cow, horse, dog and rhesus monkey.
Gene Accession No. Q9BVC4
Monoclonal or Polyclonal Polyclonal
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Concentration 0.5mg/mL
Primary or Secondary Primary
Gene ID (Entrez) 64223
Conjugate Unlabeled
Quantity 0.025mg
Immunogen A portion of amino acids 150-200 of human LST8 was used as the immunogen.
Applications Western Blotting
Format Purified
Host Species Rabbit
Antigen GBL
Regulatory Status RUO
Isotype IgG