missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-GATA-4, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 330-380 of human GATA-4 was used as the immunogen for the antibody.
Antigen GATA-4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224585SS Novus Biologicals Canada
NBP224585SS
0.025mg Each for $109.00
NBP224585 Novus Biologicals Canada
NBP224585
0.1mg Each for $257.41
Specifications Description & Specifications

Specifications

Rabbit
GATA-4
Affinity Purified
RUO
The amino acid sequence used as immunogen for the HSP27 antibody is 100% homologous in human, horse, macaque, chimpanzee and dog, 90% homologous in opossum, cow, zebrafish, boar and Xenopus and 81% homologous in rat and mouse.
2626
0.5mg/mL
A portion of amino acids 330-380 of human GATA-4 was used as the immunogen for the antibody.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
P43694
100μg in 200μL PBS containing 0.05% BSA