Please login to your online account to display your discounted pricing

anti-Gastrokine 1, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Isotype IgG
Immunogen The portions of amino acids range between 100-150 of human GKN1 was used as the immunogen.
Antigen Gastrokine 1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224766SS

 
novus biologicals canada
NBP224766SS
Unlabeled 0.025mg Each for $139.00

NBP224766

 
novus biologicals canada
NBP224766
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen The portions of amino acids range between 100-150 of human GKN1 was used as the immunogen.
Antigen Gastrokine 1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and dog, 94% homologous in mouse and cow, 88% homologous in rat and 77% homologous in chicken.
Gene ID (Entrez) 56287
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL