Please login to your online account to display your discounted pricing

anti-Gastric Lipase, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within residues 1-100 of human Gastric Lipase. [Swiss-Prot No. P07098]
Antigen Gastric Lipase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11060930

 
novus biologicals canada
NB11060930
Unlabeled 0.1mL Each for $309.05

NB11060930T

 
novus biologicals canada
NB11060930SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within residues 1-100 of human Gastric Lipase. [Swiss-Prot No. P07098]
Antigen Gastric Lipase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Human and mouse. Immunogen sequence has 87% homology to canine.
Gene Accession No. P07098
Gene ID (Entrez) 8513
Formulation PBS, 30% glycerol
Concentration 0.9mg/mL