Please login to your online account to display your discounted pricing

anti-GAPDH, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300327SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300327SS

  • / Each
 

Description & Specifications

Specifications

Antigen GAPDH
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Avian
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Invertebrate
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Yeast
Format Antisera
Host Species Rabbit
Immunogen Full length GAPDH purified from bovine brain. [UniProt No. P10096]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 2597
Test Specificity Human, cow, pig, mouse, rat, bird and chicken. Yeast reactivity reported in scientific literature (PMID: 23709183). Since GAPDH is one of the most conserved proteins known and it is therefore likely that the antibody will react with other species also. Invertebrate reactivity reported in scientific literature (PMID: 18638509).
Target Species Avian
Target Species Bovine
Target Species Chicken
Target Species Human
Target Species Invertebrate
Target Species Murine
Target Species Porcine
Target Species Rat
Target Species Yeast