Please login to your online account to display your discounted pricing

anti-GAPDH, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300326SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300326SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 2597
Antigen GAPDH
Immunogen The epitope recognized by this antibody maps to a region between residues 300 and the C-terminus (residue 335) of mouse GAPDH using the numbering given in entry NP_001001303.1 (GeneID 407972).
Test Specificity Mouse. In tests of lysate from HeLa cells, reactivity with Human GAPDH was weak.
Conjugate Unlabeled
Formulation TBS with 0.1% BSA.
Applications Simple Western
Applications Western Blotting
Concentration 0.2mg/mL
Host Species Rabbit
Cross Reactivity Mouse
Format Affinity Purified
Quantity 0.025mL
Regulatory Status RUO