Please login to your online account to display your discounted pricing

anti-GAPDH, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300323SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300323SS

  • / Each
 

Description & Specifications

Specifications

Concentration 0.2mg/mL
Gene ID (Entrez) 2597
Monoclonal or Polyclonal Polyclonal
Gene Accession No. P04406
Formulation TBS with 0.1% BSA.
Immunogen The epitope recognized by this antibody maps to a region between residues 200 and 250 of human GAPDH using the numbering given in entry NP_002037.2 (GeneID 2597).
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Xenopus
Cross Reactivity Yeast
Conjugate Unlabeled
Test Specificity Human and mouse. Reactivity with X. laevis and S. Pombe and S. cerevisae GAPDH is expected owing to conservation of sequence between this ortholog and Human GAPDH at the site to which the epitope maps.
Quantity 0.025mL
Applications Simple Western
Applications Western Blotting
Host Species Rabbit
Antigen GAPDH
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO