Please login to your online account to display your discounted pricing

anti-GAPDH, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300322SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300322SS

  • / Each
 

Description & Specifications

Specifications

Concentration 0.2mg/mL
Gene Accession No. P04406
Gene ID (Entrez) 2597
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Primary or Secondary Primary
Test Specificity Reacts with Human, Mouse, Rat and Primate sp. Based upon 100% sequence homology, this antibody is expected to react with Hamster (Chinese hamster, Golden Hamster), Canines, Ferret, Squirrel, Pig/Porcine, Bovine, Sheep, Rabbit, Xenopus and other amphibians such as Turtle, Avian species (including Chicken, Turkey, Sparrows, Duck), Zebrafish and several other fishes.
Monoclonal or Polyclonal Polyclonal
Formulation TBS with 0.1% BSA.
Quantity 0.025mL
Antigen GAPDH
Immunogen The epitope recognized by this antibody maps to a region between residues 150 and 200 of human GAPDH using the numbering given in entry NP_002037.2 (GeneID 2597).
Regulatory Status RUO
Format Affinity Purified
Host Species Rabbit