Please login to your online account to display your discounted pricing

anti-GAPDH, Polyclonal, Novus Biologicals™

$111.78 - $307.00

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residues 200 and 250 of human GAPDH using the numbering given in entry NP_002037.2 (GeneID 2597).
Antigen GAPDH
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB300323

 
novus biologicals canada
NB300323
Unlabeled 0.1mL Each for $307.00

NB300323SS

 
novus biologicals canada
NB300323SS
Unlabeled 0.025mL Each for $111.78
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The epitope recognized by this antibody maps to a region between residues 200 and 250 of human GAPDH using the numbering given in entry NP_002037.2 (GeneID 2597).
Antigen GAPDH
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Simple Western
Applications Western Blot
Test Specificity Human and mouse. Reactivity with X. laevis and S. Pombe and S. cerevisae GAPDH is expected owing to conservation of sequence between this ortholog and Human GAPDH at the site to which the epitope maps.
Gene Accession No. P04406
Gene ID (Entrez) 2597
Formulation TBS with 0.1% BSA.
Concentration 0.2mg/mL