Please login to your online account to display your discounted pricing

anti-GAP-43, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Sheep
Immunogen GAP43 from rat brain [UniProt No. P07936]
Antigen GAP-43
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP141123

 
novus biologicals canada
NBP141123
Unlabeled 0.1mL Each for $329.00

NBP141123SS

 
novus biologicals canada
NBP141123SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Sheep
Immunogen GAP43 from rat brain [UniProt No. P07936]
Antigen GAP-43
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene ID (Entrez) 2596
Formulation PBS