Please login to your online account to display your discounted pricing

anti-GAD1/GAD67, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056385SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056385T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_000808
Gene ID (Entrez) 2571
Concentration 0.5mg/mL
Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Regulatory Status RUO
Test Specificity Reacts with Human and mouse. 94% sequence identity with rat protein.
Antigen GAD1/GAD67
Primary or Secondary Primary
Cross Reactivity Human
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of amino acids 1-100 of human GAD67. The sequence used is 100% homologous with mouse and 94% homologous with rat GAD67.
Format Purified
Applications Western Blotting
Quantity 0.025mg
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit