Please login to your online account to display your discounted pricing

anti-GAD1/GAD67, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056385

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056385

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Gene Accession No. NP_000808
Test Specificity Reacts with Human and mouse. 94% sequence identity with rat protein.
Gene ID (Entrez) 2571
Conjugate Unlabeled
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Formulation PBS containing 0.05% BSA
Regulatory Status RUO
Format Purified
Host Species Rabbit
Antigen GAD1/GAD67
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of amino acids 1-100 of human GAD67. The sequence used is 100% homologous with mouse and 94% homologous with rat GAD67.
Quantity 0.1mg
Cross Reactivity Human