Please login to your online account to display your discounted pricing

anti-GAD1/GAD67, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of amino acids 1-100 of human GAD67. The sequence used is 100% homologous with mouse and 94% homologous with rat GAD67.
Antigen GAD1/GAD67
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056385

 
novus biologicals canada
NB10056385
Unlabeled 0.1mg Each for $255.80

NB10056385T

 
novus biologicals canada
NB10056385SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of amino acids 1-100 of human GAD67. The sequence used is 100% homologous with mouse and 94% homologous with rat GAD67.
Antigen GAD1/GAD67
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Reacts with Human and mouse. 94% sequence identity with rat protein.
Gene Accession No. NP_000808
Gene ID (Entrez) 2571
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL